| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010834.hg.1 |
ENST00000505861.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010834.hg.1 |
ENST00000505861.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010834.hg.1 |
ENST00000505861.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010834.hg.1 |
ENST00000505861.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010834.hg.1 |
NR_104669.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010834.hg.1 |
NR_104669.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010834.hg.1 |
NR_104669.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010834.hg.1 |
NR_104669.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500010834.hg.1 |
NR_104669.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500175519.hg.1 |
ENST00000505861.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500175519.hg.1 |
NR_104669.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500175519.hg.1 |
ENST00000505861.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500175519.hg.1 |
NR_104669.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500175519.hg.1 |
TC0500010834.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500175521.hg.1 |
ENST00000505861.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500175521.hg.1 |
NR_104669.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500175521.hg.1 |
ENST00000505861.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500175521.hg.1 |
NR_104669.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500175521.hg.1 |
TC0500010834.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500010834.hg.1 |
ENST00000505861.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010834.hg.1 |
ENST00000505861.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010834.hg.1 |
ENST00000505861.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010834.hg.1 |
ENST00000505861.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500010834.hg.1 |
NR_104669.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010834.hg.1 |
NR_104669.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010834.hg.1 |
NR_104669.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010834.hg.1 |
NR_104669.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500010834.hg.1 |
NR_104669.1 |
Refseq |