Assay Details
| Gene Symbol: TPD52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_105033.1 | 4 - 5 | 853 | 93 | ||
| GenBank mRNA | AK057075.1 | - | 5 - 6 | 571 | 93 | |
| GQ499328.1 | - | 4 - 5 | 838 | 93 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitochondrial ribosomal protein S28 | MRPS28 |
HSPC007, MRP-S28, MRP-S35, MRPS35 |
-21.3kb | Sense | Chr. 8 - 79918860 - 80030271 on NCBI Build GRCh38 |
| Downstream | microRNA 5708 | MIR5708 | 188.7kb | Anti-sense | Chr. 8 - 80241389 - 80241473 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

TPD52
Genomic Map