Assay Details
| Gene Symbol: MIR6076 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106724.1 | 1 | 59 | 94 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100506446 | LOC100506446 | -4.4kb | Anti-sense | Chr. 14 - 49919826 - 49963124 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 1588 | LINC01588 | C14orf182 | 15.2kb | Anti-sense | Chr. 14 - 49981669 - 50007520 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1400007102.hg.1
Set Membership: |
|

MIR6076
Genomic Map