Assay Details
| Gene Symbol: MIR6129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106745.1 | 1 | 51 | 88 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized FLJ40194 | FLJ40194 | -29.7kb | Anti-sense | Chr. 17 - 49248243 - 49258665 on NCBI Build GRCh38 | |
| Downstream | zinc finger protein 652 | ZNF652 | 752.0 | Sense | Chr. 17 - 49289206 - 49362473 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1700011037.hg.1
Set Membership: |
|

MIR6129
Genomic Map