Assay Details
| Gene Symbol: MIR6507 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106762.1 | 1 | 36 | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101927278 | LOC101927278 | -470.8kb | Anti-sense | Chr. 10 - 98446321 - 98453805 on NCBI Build GRCh38 | |
| Downstream | cyclin and CBS domain divalent metal cation transport mediator 1 | CNNM1 |
ACDP1, CLP-1 |
404.5kb | Anti-sense | Chr. 10 - 99329099 - 99394330 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1000011582.hg.1
Set Membership: |
|

MIR6507
Genomic Map