Assay Details
| Gene Symbol: MIR6718 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106776.1 | 1 | 43 | 65 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105371967 | LOC105371967 | -229.1kb | Sense | Chr. 18 - 3653056 - 3656282 on NCBI Build GRCh38 | |
| Downstream | DLGAP1 antisense RNA 4 | DLGAP1-AS4 | 76.9kb | Sense | Chr. 18 - 3962353 - 4013943 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1800006529.hg.1
Set Membership: |
|

MIR6718
Genomic Map