Assay Details
| Gene Symbol: MIR6856 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106915.1 | 1 | 35 | 67 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100272217 | LOC100272217 | -46.8kb | Anti-sense | Chr. 9 - 130577350 - 130579494 on NCBI Build GRCh38 | |
| Downstream | PR domain 12 | PRDM12 |
HSAN8, PFM9 |
38.2kb | Sense | Chr. 9 - 130664594 - 130682997 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0900008962.hg.1
Set Membership: |
|

MIR6856
Genomic Map