Assay Details
| Gene Symbol: MIR6871 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106931.1 | 1 | 26 | 51 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PLCG1 antisense RNA 1 | PLCG1-AS1 | TOP1-AS1 | -31.0kb | Anti-sense | Chr. 20 - 41097994 - 41138000 on NCBI Build GRCh38 |
| Downstream | zinc fingers and homeoboxes 3 | ZHX3 | TIX1 | 9.4kb | Anti-sense | Chr. 20 - 41178449 - 41317606 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC2000007389.hg.1
Set Membership: |
|

MIR6871
Genomic Map