Assay Details
| Gene Symbol: MIR6892 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106952.1 | 1 | 58 | 95 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | family with sequence similarity 131 member B | FAM131B | -382.0 | Anti-sense | Chr. 7 - 143353400 - 143382304 on NCBI Build GRCh38 | |
| Downstream | EPH receptor A1 | EPHA1 |
EPH, EPHT, EPHT1 |
8.0kb | Anti-sense | Chr. 7 - 143390813 - 143408892 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0700009494.hg.1
Set Membership: |
|

MIR6892
Genomic Map