Assay Details
| Gene Symbol: MIR7702 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106989.1 | 1 | 27 | 59 | ||
| GenBank mRNA | KC832799.1 | - | 1 | 27 | 59 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | lysophosphatidic acid receptor 1 | LPAR1 |
EDG2, GPR26, Gpcr26, LPA1, Mrec1.3, VZG1, edg-2, rec.1.3, vzg-1 |
-231.9kb | Sense | Chr. 9 - 110873252 - 111039259 on NCBI Build GRCh38 |
| Downstream | olfactory receptor family 2 subfamily K member 2 | OR2K2 |
HSHTPCRH06, HTPCRH06, OR2AN1P, OR2AR1P |
56.2kb | Sense | Chr. 9 - 111326884 - 111330224 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0900011197.hg.1
Set Membership: |
|

MIR7702
Genomic Map