Assay Details
| Gene Symbol: MIR8082 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_107049.1 | 1 | 44 | 70 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 1243 | MIR1243 |
MIRN1243, hsa-mir-1243 |
-45.3kb | Sense | Chr. 4 - 113106863 - 113106955 on NCBI Build GRCh38 |
| Downstream | ribosomal protein S26 pseudogene 25 | RPS26P25 | RPS26_10_512 | 61.6kb | Anti-sense | Chr. 4 - 113213969 - 113214420 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0400008454.hg.1
Set Membership: |
|

MIR8082
Genomic Map