| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012784.hg.1 |
ENST00000506431.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012784.hg.1 |
ENST00000506431.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012784.hg.1 |
ENST00000506431.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012784.hg.1 |
ENST00000506431.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012784.hg.1 |
ENST00000506431.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012784.hg.1 |
ENST00000506431.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012784.hg.1 |
ENST00000506431.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012784.hg.1 |
ENST00000506431.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012784.hg.1 |
ENST00000506431.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012784.hg.1 |
ENST00000506431.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012784.hg.1 |
ENST00000506431.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012784.hg.1 |
ENST00000520275.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012784.hg.1 |
ENST00000520275.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012784.hg.1 |
ENST00000520275.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012784.hg.1 |
NR_108022.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012784.hg.1 |
NR_108022.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012784.hg.1 |
NR_108022.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012784.hg.1 |
NR_108022.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012784.hg.1 |
NR_108022.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012784.hg.1 |
NR_108022.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012784.hg.1 |
NR_108022.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012784.hg.1 |
NR_108022.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500188975.hg.1 |
ENST00000506431.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500188975.hg.1 |
NR_108022.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500188975.hg.1 |
ENST00000506431.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500188975.hg.1 |
NR_108022.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500188975.hg.1 |
TC0500012784.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500012784.hg.1 |
ENST00000506431.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012784.hg.1 |
ENST00000506431.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012784.hg.1 |
ENST00000506431.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012784.hg.1 |
ENST00000506431.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012784.hg.1 |
ENST00000506431.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012784.hg.1 |
ENST00000506431.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012784.hg.1 |
ENST00000506431.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012784.hg.1 |
ENST00000506431.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012784.hg.1 |
ENST00000506431.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012784.hg.1 |
ENST00000506431.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012784.hg.1 |
ENST00000506431.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012784.hg.1 |
ENST00000520275.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012784.hg.1 |
ENST00000520275.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012784.hg.1 |
ENST00000520275.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012784.hg.1 |
NR_108022.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012784.hg.1 |
NR_108022.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012784.hg.1 |
NR_108022.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012784.hg.1 |
NR_108022.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012784.hg.1 |
NR_108022.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012784.hg.1 |
NR_108022.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012784.hg.1 |
NR_108022.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012784.hg.1 |
NR_108022.1 |
Refseq |