| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012040.hg.1 |
ENST00000508936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012040.hg.1 |
ENST00000508936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012040.hg.1 |
NR_108077.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012040.hg.1 |
NR_108077.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012040.hg.1 |
ENST00000508936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012040.hg.1 |
ENST00000508936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012040.hg.1 |
NR_108077.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012040.hg.1 |
NR_108077.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400182646.hg.1 |
ENST00000508936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400182646.hg.1 |
NR_108077.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400182646.hg.1 |
ENST00000508936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400182646.hg.1 |
NR_108077.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400182646.hg.1 |
TC0400012040.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400182647.hg.1 |
ENST00000508936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400182647.hg.1 |
NR_108077.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400182647.hg.1 |
ENST00000508936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400182647.hg.1 |
NR_108077.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400182647.hg.1 |
TC0400012040.hg.1 |
Clariom D TC |