Assay Details
| Gene Symbol: NR2F1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_021491.2 | 5 - 6 | 545 | 82 | ||
| NR_109821.1 | 6 - 7 | 666 | 82 | |||
| GenBank mRNA | AK311143.1 | - | 5 - 6 | 444 | 82 | |
| BC042879.1 | - | 6 - 7 | 565 | 82 | ||
| BX647510.1 | - | 5 - 6 | 360 | 82 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ARRDC3 antisense RNA 1 | ARRDC3-AS1 | -2.0mb | Anti-sense | Chr. 5 - 91380347 - 91420715 on NCBI Build GRCh38 | |
| Downstream | nuclear receptor subfamily 2 group F member 1 | NR2F1 |
BBOAS, BBSOAS, COUP-TFI, EAR-3, EAR3, ERBAL3, NR2F2, SVP44, TCFCOUP1, TFCOUP1 |
144.6kb | Anti-sense | Chr. 5 - 93583337 - 93594613 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

NR2F1-AS1
Genomic Map