| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
ENST00000507558.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
ENST00000507558.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
ENST00000508424.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
ENST00000508424.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
ENST00000508424.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
ENST00000508424.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
ENST00000508424.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
ENST00000508424.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
ENST00000508424.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
ENST00000508424.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
ENST00000508424.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
ENST00000508424.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
ENST00000508424.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
ENST00000508424.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
ENST00000508640.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
ENST00000508640.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
ENST00000508640.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
ENST00000508640.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
ENST00000510682.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
ENST00000510682.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
ENST00000510682.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
ENST00000510682.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
ENST00000510682.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
ENST00000512128.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
ENST00000512128.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
ENST00000512128.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
ENST00000512128.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
ENST00000512128.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
ENST00000512128.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
ENST00000514214.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
ENST00000514214.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
ENST00000514214.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
NR_109879.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
NR_109879.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
NR_109879.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
NR_109879.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
NR_109879.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
NR_109879.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
NR_109879.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008402.hg.1 |
NR_109879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
ENST00000507558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
ENST00000507558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
ENST00000508424.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
ENST00000508424.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
ENST00000508424.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
ENST00000508424.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
ENST00000508424.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
ENST00000508424.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
ENST00000508424.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
ENST00000508424.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
ENST00000508424.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
ENST00000508424.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
ENST00000508424.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
ENST00000508424.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
ENST00000508640.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
ENST00000508640.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
ENST00000508640.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
ENST00000508640.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
ENST00000510682.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
ENST00000510682.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
ENST00000510682.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
ENST00000510682.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
ENST00000510682.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
ENST00000512128.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
ENST00000512128.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
ENST00000512128.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
ENST00000512128.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
ENST00000512128.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
ENST00000512128.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
ENST00000514214.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
ENST00000514214.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
ENST00000514214.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
NR_109879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
NR_109879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
NR_109879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
NR_109879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
NR_109879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
NR_109879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
NR_109879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008402.hg.1 |
NR_109879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500157539.hg.1 |
ENST00000508424.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157539.hg.1 |
ENST00000508424.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157539.hg.1 |
ENST00000508640.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157539.hg.1 |
ENST00000510682.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157539.hg.1 |
ENST00000512128.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157539.hg.1 |
ENST00000514214.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157539.hg.1 |
NR_109879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500157539.hg.1 |
ENST00000508424.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157539.hg.1 |
ENST00000508424.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157539.hg.1 |
ENST00000508640.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157539.hg.1 |
ENST00000510682.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157539.hg.1 |
ENST00000512128.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157539.hg.1 |
ENST00000514214.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157539.hg.1 |
NR_109879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500157539.hg.1 |
TC0500008402.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500157538.hg.1 |
ENST00000508424.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157538.hg.1 |
ENST00000508424.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157538.hg.1 |
ENST00000508640.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157538.hg.1 |
ENST00000510682.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157538.hg.1 |
ENST00000512128.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157538.hg.1 |
NR_109879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500157538.hg.1 |
ENST00000508424.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157538.hg.1 |
ENST00000508424.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157538.hg.1 |
ENST00000508640.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157538.hg.1 |
ENST00000510682.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157538.hg.1 |
ENST00000512128.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157538.hg.1 |
NR_109879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500157538.hg.1 |
TC0500008402.hg.1 |
Clariom D TC |