Assay Details
| Gene Symbol: SEMA6A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_109879.1 | 2 - 3 | 482 | 88 | ||
| GenBank mRNA | CD300011.1 | - | 2 - 3 | 377 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | semaphorin 6A | SEMA6A |
HT018, SEMA, SEMA6A1, SEMAQ, VIA |
Anti-sense | Chr. 5 - 116443555 - 116574926 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC102467223 | LOC102467223 | 271.6kb | Anti-sense | Chr. 5 - 116743302 - 116762209 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000507558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000507558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000508640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000508640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000508640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000508640.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000510682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000510682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000510682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000510682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000510682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000512128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000512128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000512128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000512128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000512128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000512128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000514214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000514214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | ENST00000514214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008402.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000507558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000507558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000508640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000508640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000508640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000508640.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000510682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000510682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000510682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000510682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000510682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000512128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000512128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000512128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000512128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000512128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000512128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000514214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000514214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | ENST00000514214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008402.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157542.hg.1 | ENST00000507558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157542.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157542.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157542.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157542.hg.1 | ENST00000507558.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157542.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157542.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157542.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157542.hg.1 | TC0500008402.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500157543.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157543.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157543.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157543.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157543.hg.1 | ENST00000508424.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157543.hg.1 | NR_109879.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157543.hg.1 | TC0500008402.hg.1 | Clariom D TC |

SEMA6A-AS1
Genomic Map