| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009126.hg.1 |
ENST00000454626.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009126.hg.1 |
ENST00000454626.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009126.hg.1 |
ENST00000454626.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009126.hg.1 |
ENST00000454626.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009126.hg.1 |
ENST00000454626.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009126.hg.1 |
ENST00000454626.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009126.hg.1 |
ENST00000454626.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009126.hg.1 |
ENST00000454626.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009126.hg.1 |
NR_109889.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009126.hg.1 |
NR_109889.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009126.hg.1 |
NR_109889.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009126.hg.1 |
NR_109889.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009126.hg.1 |
NR_109889.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009126.hg.1 |
NR_109889.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009126.hg.1 |
NR_109889.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009126.hg.1 |
NR_109889.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009126.hg.1 |
NR_109889.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009126.hg.1 |
NR_109889.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000163868.hg.1 |
ENST00000454626.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000163868.hg.1 |
NR_109889.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000163868.hg.1 |
TC2000009126.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000163868.hg.1 |
ENST00000454626.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000163868.hg.1 |
NR_109889.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000163869.hg.1 |
ENST00000454626.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000163869.hg.1 |
NR_109889.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000163869.hg.1 |
TC2000009126.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000163869.hg.1 |
ENST00000454626.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000163869.hg.1 |
NR_109889.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000163870.hg.1 |
ENST00000454626.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000163870.hg.1 |
NR_109889.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000163870.hg.1 |
TC2000009126.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000163870.hg.1 |
ENST00000454626.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000163870.hg.1 |
NR_109889.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000163866.hg.1 |
NR_109889.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000163866.hg.1 |
TC2000009126.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000163866.hg.1 |
NR_109889.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000163867.hg.1 |
NR_109889.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000163867.hg.1 |
TC2000009126.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000163867.hg.1 |
NR_109889.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009126.hg.1 |
ENST00000454626.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009126.hg.1 |
ENST00000454626.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009126.hg.1 |
ENST00000454626.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009126.hg.1 |
ENST00000454626.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009126.hg.1 |
ENST00000454626.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009126.hg.1 |
ENST00000454626.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009126.hg.1 |
ENST00000454626.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009126.hg.1 |
ENST00000454626.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009126.hg.1 |
NR_109889.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009126.hg.1 |
NR_109889.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009126.hg.1 |
NR_109889.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009126.hg.1 |
NR_109889.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009126.hg.1 |
NR_109889.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009126.hg.1 |
NR_109889.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009126.hg.1 |
NR_109889.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009126.hg.1 |
NR_109889.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009126.hg.1 |
NR_109889.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009126.hg.1 |
NR_109889.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000163873.hg.1 |
ENST00000454626.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000163873.hg.1 |
NR_109889.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000163873.hg.1 |
TC2000009126.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000163873.hg.1 |
ENST00000454626.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000163873.hg.1 |
NR_109889.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000163871.hg.1 |
ENST00000454626.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000163871.hg.1 |
NR_109889.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000163871.hg.1 |
TC2000009126.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000163871.hg.1 |
ENST00000454626.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000163871.hg.1 |
NR_109889.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000163872.hg.1 |
ENST00000454626.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000163872.hg.1 |
NR_109889.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000163872.hg.1 |
TC2000009126.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000163872.hg.1 |
ENST00000454626.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000163872.hg.1 |
NR_109889.1 |
Refseq |