Assay Details
| Gene Symbol: LOC728095 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_109897.1 | 2 - 3 | 240 | 86 | ||
| GenBank mRNA | DB028580.1 | - | 2 - 3 | 242 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 218-2 | MIR218-2 |
MIRN218-2, mir-218-2 |
-258.1kb | Anti-sense | Chr. 5 - 168768146 - 168768255 on NCBI Build GRCh38 |
| Downstream | microRNA 585 | MIR585 |
MIRN585, hsa-mir-585, mir-585 |
233.1kb | Anti-sense | Chr. 5 - 169263601 - 169263694 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000507092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000507092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000507092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000507092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000507092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000507092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000507092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000507092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000507304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000507304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000507304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000507304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000507304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000507304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000507304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000508470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000508470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000508600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000508600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000508600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000508600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000508600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000509170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000509170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000509170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000509170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000509170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | NR_109897.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | NR_109897.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | NR_109897.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | NR_109897.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009370.hg.1 | NR_109897.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000504527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000507092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000507092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000507092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000507092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000507092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000507092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000507092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000507092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000507304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000507304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000507304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000507304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000507304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000507304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000507304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000508470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000508470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000508547.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000508600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000508600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000508600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000508600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000508600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000509170.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000509170.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000509170.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000509170.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000509170.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000511447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000512417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | ENST00000513790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | NR_109897.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | NR_109897.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | NR_109897.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | NR_109897.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009370.hg.1 | NR_109897.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500165586.hg.1 | ENST00000509170.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500165586.hg.1 | NR_109897.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500165586.hg.1 | ENST00000509170.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500165586.hg.1 | NR_109897.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500165586.hg.1 | TC0500009370.hg.1 | Clariom D TC |

LOC728095
Genomic Map