Assay Details
| Gene Symbol: LAMA5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_109922.1 | 1 - 2 | 325 | 71 | ||
| GenBank mRNA | BM700151.1 | - | 1 - 2 | 71 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 4758 | MIR4758 | -20.5kb | Anti-sense | Chr. 20 - 62332487 - 62332557 on NCBI Build GRCh38 | |
| Downstream | ribosomal protein S21 | RPS21 |
HLDF, S21 |
31.6kb | Sense | Chr. 20 - 62387065 - 62388520 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000008004.hg.1 | ENST00000456721.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008004.hg.1 | ENST00000456721.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008004.hg.1 | ENST00000456721.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008004.hg.1 | ENST00000477848.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008004.hg.1 | ENST00000477848.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008004.hg.1 | ENST00000477848.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008004.hg.1 | ENST00000477848.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008004.hg.1 | ENST00000477848.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008004.hg.1 | ENST00000477848.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008004.hg.1 | ENST00000477848.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008004.hg.1 | ENST00000477848.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008004.hg.1 | ENST00000477848.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008004.hg.1 | ENST00000477848.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008004.hg.1 | ENST00000478167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008004.hg.1 | ENST00000478167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008004.hg.1 | ENST00000478167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008004.hg.1 | ENST00000478167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008004.hg.1 | ENST00000478167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008004.hg.1 | ENST00000487421.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008004.hg.1 | ENST00000487421.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008004.hg.1 | ENST00000487421.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008004.hg.1 | ENST00000487421.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008004.hg.1 | NR_109922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008004.hg.1 | NR_109922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008004.hg.1 | NR_109922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008004.hg.1 | NR_109922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008004.hg.1 | NR_109922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008004.hg.1 | NR_109922.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008004.hg.1 | NR_109923.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008004.hg.1 | NR_109923.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008004.hg.1 | NR_109923.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008004.hg.1 | NR_109924.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008004.hg.1 | NR_109924.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008004.hg.1 | NR_109924.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008004.hg.1 | NR_109924.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008004.hg.1 | NR_109924.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008004.hg.1 | ENST00000456721.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008004.hg.1 | ENST00000456721.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008004.hg.1 | ENST00000456721.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008004.hg.1 | ENST00000477848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008004.hg.1 | ENST00000477848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008004.hg.1 | ENST00000477848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008004.hg.1 | ENST00000477848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008004.hg.1 | ENST00000477848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008004.hg.1 | ENST00000477848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008004.hg.1 | ENST00000477848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008004.hg.1 | ENST00000477848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008004.hg.1 | ENST00000477848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008004.hg.1 | ENST00000477848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008004.hg.1 | ENST00000478167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008004.hg.1 | ENST00000478167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008004.hg.1 | ENST00000478167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008004.hg.1 | ENST00000478167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008004.hg.1 | ENST00000478167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008004.hg.1 | ENST00000487421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008004.hg.1 | ENST00000487421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008004.hg.1 | ENST00000487421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008004.hg.1 | ENST00000487421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008004.hg.1 | NR_109922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008004.hg.1 | NR_109922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008004.hg.1 | NR_109922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008004.hg.1 | NR_109922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008004.hg.1 | NR_109922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008004.hg.1 | NR_109922.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008004.hg.1 | NR_109923.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008004.hg.1 | NR_109923.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008004.hg.1 | NR_109923.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008004.hg.1 | NR_109924.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008004.hg.1 | NR_109924.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008004.hg.1 | NR_109924.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008004.hg.1 | NR_109924.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008004.hg.1 | NR_109924.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000155707.hg.1 | ENST00000477848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000155707.hg.1 | ENST00000477848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000155707.hg.1 | ENST00000478167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000155707.hg.1 | NR_109922.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000155707.hg.1 | NR_109924.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000155707.hg.1 | TC2000008004.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000155707.hg.1 | ENST00000477848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000155707.hg.1 | ENST00000477848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000155707.hg.1 | ENST00000478167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000155707.hg.1 | NR_109922.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000155707.hg.1 | NR_109924.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000155705.hg.1 | ENST00000477848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000155705.hg.1 | ENST00000477848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000155705.hg.1 | ENST00000487421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000155705.hg.1 | NR_109922.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000155705.hg.1 | TC2000008004.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000155705.hg.1 | ENST00000477848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000155705.hg.1 | ENST00000477848.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000155705.hg.1 | ENST00000487421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000155705.hg.1 | NR_109922.1 | Refseq |

LAMA5-AS1
Genomic Map