| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008004.hg.1 |
ENST00000456721.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008004.hg.1 |
ENST00000456721.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008004.hg.1 |
ENST00000456721.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008004.hg.1 |
ENST00000477848.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008004.hg.1 |
ENST00000477848.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008004.hg.1 |
ENST00000477848.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008004.hg.1 |
ENST00000477848.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008004.hg.1 |
ENST00000477848.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008004.hg.1 |
ENST00000477848.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008004.hg.1 |
ENST00000477848.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008004.hg.1 |
ENST00000477848.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008004.hg.1 |
ENST00000477848.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008004.hg.1 |
ENST00000477848.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008004.hg.1 |
ENST00000478167.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008004.hg.1 |
ENST00000478167.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008004.hg.1 |
ENST00000478167.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008004.hg.1 |
ENST00000478167.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008004.hg.1 |
ENST00000478167.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008004.hg.1 |
ENST00000487421.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008004.hg.1 |
ENST00000487421.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008004.hg.1 |
ENST00000487421.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008004.hg.1 |
ENST00000487421.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008004.hg.1 |
NR_109922.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008004.hg.1 |
NR_109922.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008004.hg.1 |
NR_109922.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008004.hg.1 |
NR_109922.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008004.hg.1 |
NR_109922.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008004.hg.1 |
NR_109922.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008004.hg.1 |
NR_109923.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008004.hg.1 |
NR_109923.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008004.hg.1 |
NR_109923.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008004.hg.1 |
NR_109924.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008004.hg.1 |
NR_109924.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008004.hg.1 |
NR_109924.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008004.hg.1 |
NR_109924.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008004.hg.1 |
NR_109924.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000155703.hg.1 |
ENST00000456721.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000155703.hg.1 |
ENST00000477848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000155703.hg.1 |
ENST00000477848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000155703.hg.1 |
ENST00000478167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000155703.hg.1 |
ENST00000487421.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000155703.hg.1 |
NR_109922.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000155703.hg.1 |
NR_109923.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000155703.hg.1 |
NR_109924.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000155703.hg.1 |
TC2000008004.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000155703.hg.1 |
ENST00000456721.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000155703.hg.1 |
ENST00000477848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000155703.hg.1 |
ENST00000477848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000155703.hg.1 |
ENST00000478167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000155703.hg.1 |
ENST00000487421.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000155703.hg.1 |
NR_109922.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000155703.hg.1 |
NR_109923.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000155703.hg.1 |
NR_109924.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000155710.hg.1 |
ENST00000456721.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000155710.hg.1 |
ENST00000478167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000155710.hg.1 |
ENST00000487421.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000155710.hg.1 |
NR_109922.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000155710.hg.1 |
NR_109923.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000155710.hg.1 |
NR_109924.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000155710.hg.1 |
TC2000008004.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000155710.hg.1 |
ENST00000456721.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000155710.hg.1 |
ENST00000478167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000155710.hg.1 |
ENST00000487421.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000155710.hg.1 |
NR_109922.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000155710.hg.1 |
NR_109923.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000155710.hg.1 |
NR_109924.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000155704.hg.1 |
ENST00000456721.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000155704.hg.1 |
ENST00000477848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000155704.hg.1 |
ENST00000477848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000155704.hg.1 |
ENST00000478167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000155704.hg.1 |
ENST00000487421.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000155704.hg.1 |
NR_109922.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000155704.hg.1 |
NR_109923.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000155704.hg.1 |
NR_109924.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000155704.hg.1 |
TC2000008004.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000155704.hg.1 |
ENST00000456721.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000155704.hg.1 |
ENST00000477848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000155704.hg.1 |
ENST00000477848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000155704.hg.1 |
ENST00000478167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000155704.hg.1 |
ENST00000487421.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000155704.hg.1 |
NR_109922.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000155704.hg.1 |
NR_109923.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000155704.hg.1 |
NR_109924.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008004.hg.1 |
ENST00000456721.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008004.hg.1 |
ENST00000456721.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008004.hg.1 |
ENST00000456721.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008004.hg.1 |
ENST00000477848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008004.hg.1 |
ENST00000477848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008004.hg.1 |
ENST00000477848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008004.hg.1 |
ENST00000477848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008004.hg.1 |
ENST00000477848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008004.hg.1 |
ENST00000477848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008004.hg.1 |
ENST00000477848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008004.hg.1 |
ENST00000477848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008004.hg.1 |
ENST00000477848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008004.hg.1 |
ENST00000477848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008004.hg.1 |
ENST00000478167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008004.hg.1 |
ENST00000478167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008004.hg.1 |
ENST00000478167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008004.hg.1 |
ENST00000478167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008004.hg.1 |
ENST00000478167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008004.hg.1 |
ENST00000487421.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008004.hg.1 |
ENST00000487421.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008004.hg.1 |
ENST00000487421.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008004.hg.1 |
ENST00000487421.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008004.hg.1 |
NR_109922.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008004.hg.1 |
NR_109922.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008004.hg.1 |
NR_109922.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008004.hg.1 |
NR_109922.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008004.hg.1 |
NR_109922.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008004.hg.1 |
NR_109922.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008004.hg.1 |
NR_109923.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008004.hg.1 |
NR_109923.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008004.hg.1 |
NR_109923.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008004.hg.1 |
NR_109924.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008004.hg.1 |
NR_109924.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008004.hg.1 |
NR_109924.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008004.hg.1 |
NR_109924.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008004.hg.1 |
NR_109924.1 |
Refseq |