Assay Details
| Gene Symbol: LINC01208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_109968.1 | 1 - 2 | 91 | 117 | ||
| GenBank mRNA | BX094192.1 | - | 1 - 2 | 91 | 117 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 7977 | MIR7977 | hsa-mir-7977 | -119.2kb | Anti-sense | Chr. 3 - 176515103 - 176515151 on NCBI Build GRCh38 |
| Downstream | long intergenic non-protein coding RNA 1209 | LINC01209 | 178.6kb | Sense | Chr. 3 - 176814155 - 176817001 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LINC01208
Genomic Map