| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010211.hg.1 |
ENST00000500498.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010211.hg.1 |
ENST00000500498.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010211.hg.1 |
ENST00000500498.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010211.hg.1 |
ENST00000500498.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010211.hg.1 |
NR_109975.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010211.hg.1 |
NR_109975.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010211.hg.1 |
NR_109975.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010211.hg.1 |
NR_109975.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200177323.hg.1 |
ENST00000500498.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200177323.hg.1 |
NR_109975.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200177323.hg.1 |
ENST00000500498.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200177323.hg.1 |
NR_109975.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200177323.hg.1 |
TC1200010211.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200177324.hg.1 |
ENST00000500498.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200177324.hg.1 |
NR_109975.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200177324.hg.1 |
ENST00000500498.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200177324.hg.1 |
NR_109975.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200177324.hg.1 |
TC1200010211.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1200010211.hg.1 |
ENST00000500498.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010211.hg.1 |
ENST00000500498.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010211.hg.1 |
ENST00000500498.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010211.hg.1 |
ENST00000500498.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010211.hg.1 |
NR_109975.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010211.hg.1 |
NR_109975.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010211.hg.1 |
NR_109975.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010211.hg.1 |
NR_109975.1 |
Refseq |