Assay Details
| Gene Symbol: LOC101927058 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110042.1 | 4 - 5 | 520 | 116 | ||
| GenBank mRNA | BC042591.1 | - | 4 - 5 | 520 | 116 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | prickle planar cell polarity protein 1 | PRICKLE1 |
EPM1B, RILP |
-39.7kb | Sense | Chr. 12 - 42458338 - 42589770 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC105369738 | LOC105369738 | 12.2kb | Anti-sense | Chr. 12 - 42646583 - 42686701 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101927058
Genomic Map