| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006494.hg.1 |
ENST00000437384.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006494.hg.1 |
ENST00000437384.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006494.hg.1 |
ENST00000437384.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006494.hg.1 |
ENST00000437384.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006494.hg.1 |
ENST00000456177.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006494.hg.1 |
ENST00000456177.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006494.hg.1 |
ENST00000456177.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006494.hg.1 |
ENST00000456177.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006494.hg.1 |
ENST00000456177.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006494.hg.1 |
ENST00000456177.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006494.hg.1 |
ENST00000456177.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006494.hg.1 |
ENST00000456177.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006494.hg.1 |
ENST00000456177.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006494.hg.1 |
ENST00000456177.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006494.hg.1 |
ENST00000456177.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006494.hg.1 |
ENST00000456177.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006494.hg.1 |
NR_110090.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006494.hg.1 |
NR_110090.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006494.hg.1 |
NR_110090.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006494.hg.1 |
NR_110090.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006494.hg.1 |
NR_110090.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006494.hg.1 |
NR_110090.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000144498.hg.1 |
ENST00000456177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000144498.hg.1 |
ENST00000456177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000144498.hg.1 |
NR_110090.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000144498.hg.1 |
TC2000006494.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000144498.hg.1 |
ENST00000456177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000144498.hg.1 |
ENST00000456177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000144498.hg.1 |
NR_110090.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000144499.hg.1 |
ENST00000456177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000144499.hg.1 |
ENST00000456177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000144499.hg.1 |
NR_110090.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000144499.hg.1 |
TC2000006494.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000144499.hg.1 |
ENST00000456177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000144499.hg.1 |
ENST00000456177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000144499.hg.1 |
NR_110090.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006494.hg.1 |
ENST00000437384.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006494.hg.1 |
ENST00000437384.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006494.hg.1 |
ENST00000437384.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006494.hg.1 |
ENST00000437384.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006494.hg.1 |
ENST00000456177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006494.hg.1 |
ENST00000456177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006494.hg.1 |
ENST00000456177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006494.hg.1 |
ENST00000456177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006494.hg.1 |
ENST00000456177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006494.hg.1 |
ENST00000456177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006494.hg.1 |
ENST00000456177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006494.hg.1 |
ENST00000456177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006494.hg.1 |
ENST00000456177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006494.hg.1 |
ENST00000456177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006494.hg.1 |
ENST00000456177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006494.hg.1 |
ENST00000456177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006494.hg.1 |
NR_110090.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006494.hg.1 |
NR_110090.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006494.hg.1 |
NR_110090.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006494.hg.1 |
NR_110090.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006494.hg.1 |
NR_110090.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006494.hg.1 |
NR_110090.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000144497.hg.1 |
ENST00000456177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000144497.hg.1 |
ENST00000456177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000144497.hg.1 |
NR_110090.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000144497.hg.1 |
TC2000006494.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000144497.hg.1 |
ENST00000456177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000144497.hg.1 |
ENST00000456177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000144497.hg.1 |
NR_110090.1 |
Refseq |