| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007861.hg.1 |
ENST00000411701.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007861.hg.1 |
ENST00000411701.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007861.hg.1 |
ENST00000411701.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007861.hg.1 |
ENST00000426260.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007861.hg.1 |
ENST00000426260.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007861.hg.1 |
ENST00000426260.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007861.hg.1 |
ENST00000426260.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007861.hg.1 |
ENST00000426260.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007861.hg.1 |
ENST00000426260.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007861.hg.1 |
ENST00000426260.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007861.hg.1 |
ENST00000426260.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007861.hg.1 |
ENST00000426260.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007861.hg.1 |
ENST00000426260.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007861.hg.1 |
NR_110169.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007861.hg.1 |
NR_110169.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007861.hg.1 |
NR_110169.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007861.hg.1 |
NR_110169.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007861.hg.1 |
NR_110169.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200154266.hg.1 |
ENST00000411701.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200154266.hg.1 |
ENST00000426260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200154266.hg.1 |
ENST00000426260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200154266.hg.1 |
NR_110169.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200154266.hg.1 |
ENST00000411701.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200154266.hg.1 |
ENST00000426260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200154266.hg.1 |
ENST00000426260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200154266.hg.1 |
NR_110169.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200154266.hg.1 |
TC0200007861.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200007861.hg.1 |
ENST00000411701.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007861.hg.1 |
ENST00000411701.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007861.hg.1 |
ENST00000411701.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007861.hg.1 |
ENST00000426260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007861.hg.1 |
ENST00000426260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007861.hg.1 |
ENST00000426260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007861.hg.1 |
ENST00000426260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007861.hg.1 |
ENST00000426260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007861.hg.1 |
ENST00000426260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007861.hg.1 |
ENST00000426260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007861.hg.1 |
ENST00000426260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007861.hg.1 |
ENST00000426260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007861.hg.1 |
ENST00000426260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007861.hg.1 |
NR_110169.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007861.hg.1 |
NR_110169.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007861.hg.1 |
NR_110169.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007861.hg.1 |
NR_110169.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007861.hg.1 |
NR_110169.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200154268.hg.1 |
ENST00000426260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200154268.hg.1 |
ENST00000426260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200154268.hg.1 |
NR_110169.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200154268.hg.1 |
ENST00000426260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200154268.hg.1 |
ENST00000426260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200154268.hg.1 |
NR_110169.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200154268.hg.1 |
TC0200007861.hg.1 |
Clariom D TC |