| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006866.hg.1 |
ENST00000416575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006866.hg.1 |
ENST00000416575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006866.hg.1 |
ENST00000416575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006866.hg.1 |
ENST00000416575.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006866.hg.1 |
NR_110235.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006866.hg.1 |
NR_110235.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006866.hg.1 |
NR_110235.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006866.hg.1 |
NR_110235.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006866.hg.1 |
NR_110235.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006866.hg.1 |
NR_110235.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006866.hg.1 |
NR_110235.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200146954.hg.1 |
NR_110235.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200146954.hg.1 |
NR_110235.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200146954.hg.1 |
TC0200006866.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200146952.hg.1 |
ENST00000416575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200146952.hg.1 |
NR_110235.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200146952.hg.1 |
ENST00000416575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200146952.hg.1 |
NR_110235.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200146952.hg.1 |
TC0200006866.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200146953.hg.1 |
NR_110235.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200146953.hg.1 |
NR_110235.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200146953.hg.1 |
TC0200006866.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200146955.hg.1 |
NR_110235.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200146955.hg.1 |
NR_110235.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200146955.hg.1 |
TC0200006866.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200006866.hg.1 |
ENST00000416575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006866.hg.1 |
ENST00000416575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006866.hg.1 |
ENST00000416575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006866.hg.1 |
ENST00000416575.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006866.hg.1 |
NR_110235.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006866.hg.1 |
NR_110235.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006866.hg.1 |
NR_110235.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006866.hg.1 |
NR_110235.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006866.hg.1 |
NR_110235.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006866.hg.1 |
NR_110235.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006866.hg.1 |
NR_110235.1 |
Refseq |