| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012901.hg.1 |
ENST00000432410.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012901.hg.1 |
ENST00000432410.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012901.hg.1 |
ENST00000432410.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012901.hg.1 |
ENST00000432410.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012901.hg.1 |
ENST00000432410.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012901.hg.1 |
ENST00000432410.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012901.hg.1 |
ENST00000433432.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012901.hg.1 |
ENST00000433432.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012901.hg.1 |
ENST00000433432.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012901.hg.1 |
ENST00000433432.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012901.hg.1 |
ENST00000433432.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012901.hg.1 |
ENST00000433432.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012901.hg.1 |
ENST00000433432.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012901.hg.1 |
ENST00000451698.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012901.hg.1 |
ENST00000451698.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012901.hg.1 |
ENST00000451698.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012901.hg.1 |
ENST00000451698.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012901.hg.1 |
NR_110264.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012901.hg.1 |
NR_110264.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012901.hg.1 |
NR_110264.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012901.hg.1 |
NR_110264.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012901.hg.1 |
NR_110264.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012901.hg.1 |
NR_110264.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012901.hg.1 |
NR_110264.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012901.hg.1 |
NR_110264.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012901.hg.1 |
NR_110265.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012901.hg.1 |
NR_110265.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012901.hg.1 |
NR_110265.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012901.hg.1 |
NR_110265.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012901.hg.1 |
NR_110265.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012901.hg.1 |
NR_110265.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012901.hg.1 |
NR_110265.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012901.hg.1 |
ENST00000432410.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012901.hg.1 |
ENST00000432410.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012901.hg.1 |
ENST00000432410.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012901.hg.1 |
ENST00000432410.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012901.hg.1 |
ENST00000432410.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012901.hg.1 |
ENST00000432410.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012901.hg.1 |
ENST00000433432.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012901.hg.1 |
ENST00000433432.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012901.hg.1 |
ENST00000433432.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012901.hg.1 |
ENST00000433432.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012901.hg.1 |
ENST00000433432.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012901.hg.1 |
ENST00000433432.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012901.hg.1 |
ENST00000433432.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012901.hg.1 |
ENST00000451698.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012901.hg.1 |
ENST00000451698.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012901.hg.1 |
ENST00000451698.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012901.hg.1 |
ENST00000451698.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012901.hg.1 |
NR_110264.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012901.hg.1 |
NR_110264.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012901.hg.1 |
NR_110264.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012901.hg.1 |
NR_110264.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012901.hg.1 |
NR_110264.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012901.hg.1 |
NR_110264.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012901.hg.1 |
NR_110264.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012901.hg.1 |
NR_110264.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012901.hg.1 |
NR_110265.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012901.hg.1 |
NR_110265.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012901.hg.1 |
NR_110265.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012901.hg.1 |
NR_110265.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012901.hg.1 |
NR_110265.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012901.hg.1 |
NR_110265.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012901.hg.1 |
NR_110265.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200193866.hg.1 |
ENST00000432410.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200193866.hg.1 |
ENST00000432410.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200193866.hg.1 |
ENST00000433432.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200193866.hg.1 |
ENST00000451698.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200193866.hg.1 |
NR_110264.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200193866.hg.1 |
NR_110265.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200193866.hg.1 |
ENST00000432410.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200193866.hg.1 |
ENST00000432410.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200193866.hg.1 |
ENST00000433432.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200193866.hg.1 |
ENST00000451698.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200193866.hg.1 |
NR_110264.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200193866.hg.1 |
NR_110265.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200193866.hg.1 |
TC0200012901.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200193864.hg.1 |
ENST00000433432.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200193864.hg.1 |
ENST00000451698.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200193864.hg.1 |
NR_110264.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200193864.hg.1 |
NR_110265.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200193864.hg.1 |
ENST00000433432.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200193864.hg.1 |
ENST00000451698.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200193864.hg.1 |
NR_110264.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200193864.hg.1 |
NR_110265.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200193864.hg.1 |
TC0200012901.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200193867.hg.1 |
ENST00000433432.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200193867.hg.1 |
NR_110264.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200193867.hg.1 |
NR_110265.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200193867.hg.1 |
ENST00000433432.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200193867.hg.1 |
NR_110264.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200193867.hg.1 |
NR_110265.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200193867.hg.1 |
TC0200012901.hg.1 |
Clariom D TC |