Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR1900192269.hg.1 |
ENST00000263274.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192269.hg.1 |
ENST00000427526.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192269.hg.1 |
ENST00000536218.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192269.hg.1 |
ENST00000594067.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192269.hg.1 |
ENST00000594759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192269.hg.1 |
ENST00000601091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192269.hg.1 |
ENST00000613670.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192269.hg.1 |
NM_000234.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900192269.hg.1 |
NM_001289063.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900192269.hg.1 |
NM_001289064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900192269.hg.1 |
NR_110296.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900192269.hg.1 |
TC1900011060.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900192269.hg.1 |
ENST00000263274.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192269.hg.1 |
ENST00000427526.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192269.hg.1 |
ENST00000536218.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192269.hg.1 |
ENST00000594067.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192269.hg.1 |
ENST00000594759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192269.hg.1 |
ENST00000601091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192269.hg.1 |
ENST00000613670.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192269.hg.1 |
NM_000234.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900192269.hg.1 |
NM_001289063.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900192269.hg.1 |
NM_001289064.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900192269.hg.1 |
NR_110296.1 |
Refseq |