| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009411.hg.1 |
ENST00000554918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009411.hg.1 |
ENST00000554918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009411.hg.1 |
ENST00000554918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009411.hg.1 |
ENST00000554918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009411.hg.1 |
NR_110550.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009411.hg.1 |
NR_110550.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009411.hg.1 |
NR_110550.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009411.hg.1 |
NR_110550.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400169525.hg.1 |
ENST00000554918.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400169525.hg.1 |
NR_110550.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400169525.hg.1 |
ENST00000554918.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400169525.hg.1 |
NR_110550.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400169525.hg.1 |
TC1400009411.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1400169528.hg.1 |
ENST00000554918.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400169528.hg.1 |
NR_110550.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400169528.hg.1 |
ENST00000554918.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400169528.hg.1 |
NR_110550.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400169528.hg.1 |
TC1400009411.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1400009411.hg.1 |
ENST00000554918.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009411.hg.1 |
ENST00000554918.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009411.hg.1 |
ENST00000554918.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009411.hg.1 |
ENST00000554918.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009411.hg.1 |
NR_110550.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009411.hg.1 |
NR_110550.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009411.hg.1 |
NR_110550.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009411.hg.1 |
NR_110550.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400169527.hg.1 |
ENST00000554918.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400169527.hg.1 |
NR_110550.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400169527.hg.1 |
ENST00000554918.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400169527.hg.1 |
NR_110550.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400169527.hg.1 |
TC1400009411.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1400169526.hg.1 |
ENST00000554918.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400169526.hg.1 |
NR_110550.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400169526.hg.1 |
ENST00000554918.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400169526.hg.1 |
NR_110550.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400169526.hg.1 |
TC1400009411.hg.1 |
Clariom D TC |