| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009730.hg.1 |
ENST00000455232.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009730.hg.1 |
ENST00000455232.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009730.hg.1 |
ENST00000455232.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009730.hg.1 |
NR_110552.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009730.hg.1 |
NR_110552.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009730.hg.1 |
NR_110552.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400171831.hg.1 |
ENST00000455232.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400171831.hg.1 |
NR_110552.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400171831.hg.1 |
ENST00000455232.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400171831.hg.1 |
NR_110552.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400171831.hg.1 |
TC1400009730.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1400171832.hg.1 |
ENST00000455232.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400171832.hg.1 |
NR_110552.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400171832.hg.1 |
ENST00000455232.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400171832.hg.1 |
NR_110552.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400171832.hg.1 |
TC1400009730.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1400171833.hg.1 |
ENST00000455232.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400171833.hg.1 |
NR_110552.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400171833.hg.1 |
ENST00000455232.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400171833.hg.1 |
NR_110552.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400171833.hg.1 |
TC1400009730.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1400009730.hg.1 |
ENST00000455232.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009730.hg.1 |
ENST00000455232.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009730.hg.1 |
ENST00000455232.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009730.hg.1 |
NR_110552.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009730.hg.1 |
NR_110552.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009730.hg.1 |
NR_110552.1 |
Refseq |