| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009861.hg.1 |
ENST00000414885.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009861.hg.1 |
ENST00000414885.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009861.hg.1 |
NR_110574.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009861.hg.1 |
NR_110574.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009861.hg.1 |
NR_110574.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200168781.hg.1 |
ENST00000414885.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200168781.hg.1 |
NR_110574.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200168781.hg.1 |
ENST00000414885.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200168781.hg.1 |
NR_110574.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200168781.hg.1 |
TC0200009861.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200009861.hg.1 |
ENST00000414885.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009861.hg.1 |
ENST00000414885.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009861.hg.1 |
NR_110574.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200009861.hg.1 |
NR_110574.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200009861.hg.1 |
NR_110574.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200168782.hg.1 |
ENST00000414885.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200168782.hg.1 |
NR_110574.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200168782.hg.1 |
ENST00000414885.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200168782.hg.1 |
NR_110574.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200168782.hg.1 |
TC0200009861.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200168783.hg.1 |
NR_110574.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200168783.hg.1 |
NR_110574.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200168783.hg.1 |
TC0200009861.hg.1 |
Clariom D TC |