Assay Details
| Gene Symbol: MIR4632 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_039775.2 | 1 | 28 | 58 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | DB457812.1 | - | 1 | Chr. 1 - 12191347 - 12191824 [+] on Build GRCh38 | 58 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 7846 | MIR7846 | hsa-mir-7846 | -24.7kb | Sense | Chr. 1 - 12166943 - 12167038 on NCBI Build GRCh38 |
| Downstream | vacuolar protein sorting 13 homolog D | VPS13D | 38.3kb | Sense | Chr. 1 - 12230039 - 12512047 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0100006881.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01000155.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000489921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000489921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000489921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000489921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000536782.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000536782.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000536782.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000536782.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000536782.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000536782.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000536782.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000584158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000622420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | ENST00000622420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | NR_039775.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | NR_107000.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006881.hg.1 | NR_107000.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01002589.hg.1 | NR_039775.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01002589.hg.1 | TC01000155.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000376259.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000489921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000489921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000489921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000489921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000492361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000536782.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000536782.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000536782.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000536782.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000536782.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000536782.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000536782.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000584158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000622420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | ENST00000622420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | NM_001066.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | NR_039775.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | NR_107000.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006881.hg.1 | NR_107000.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100147929.hg.1 | ENST00000584158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100147929.hg.1 | NR_039775.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100147929.hg.1 | ENST00000584158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100147929.hg.1 | NR_039775.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100147929.hg.1 | TC0100006881.hg.1 | Clariom D TC |

MIR4632
Genomic Map