| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007780.hg.1 |
ENST00000443247.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007780.hg.1 |
ENST00000443247.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007780.hg.1 |
ENST00000443247.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007780.hg.1 |
ENST00000443247.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007780.hg.1 |
ENST00000443247.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007780.hg.1 |
ENST00000443247.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007780.hg.1 |
ENST00000443247.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007780.hg.1 |
ENST00000443247.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007780.hg.1 |
NR_110651.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007780.hg.1 |
NR_110651.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007780.hg.1 |
NR_110651.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007780.hg.1 |
NR_110651.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007780.hg.1 |
NR_110651.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007780.hg.1 |
NR_110651.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00153155.hg.1 |
ENST00000443247.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00153155.hg.1 |
NR_110651.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00153155.hg.1 |
ENST00000443247.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00153155.hg.1 |
NR_110651.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00153155.hg.1 |
TC0X00007780.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00007780.hg.1 |
ENST00000443247.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007780.hg.1 |
ENST00000443247.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007780.hg.1 |
ENST00000443247.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007780.hg.1 |
ENST00000443247.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007780.hg.1 |
ENST00000443247.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007780.hg.1 |
ENST00000443247.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007780.hg.1 |
ENST00000443247.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007780.hg.1 |
ENST00000443247.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007780.hg.1 |
NR_110651.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007780.hg.1 |
NR_110651.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007780.hg.1 |
NR_110651.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007780.hg.1 |
NR_110651.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007780.hg.1 |
NR_110651.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007780.hg.1 |
NR_110651.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00153153.hg.1 |
ENST00000443247.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00153153.hg.1 |
NR_110651.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00153153.hg.1 |
ENST00000443247.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00153153.hg.1 |
NR_110651.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00153153.hg.1 |
TC0X00007780.hg.1 |
Clariom D TC |