Assay Details
| Gene Symbol: LOC101927604 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110681.1 | 2 - 3 | 574 | 92 | ||
| GenBank mRNA | BC032911.1 | - | 2 - 3 | 579 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromosome 1 open reading frame 198 | C1orf198 | -5.7kb | Anti-sense | Chr. 1 - 230837119 - 230869589 on NCBI Build GRCh38 | |
| Downstream | tetratricopeptide repeat domain 13 | TTC13 | 27.2kb | Anti-sense | Chr. 1 - 230906241 - 230978872 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101927604
Genomic Map