| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014840.hg.1 |
ENST00000425750.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014840.hg.1 |
ENST00000425750.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014840.hg.1 |
ENST00000425750.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014840.hg.1 |
ENST00000425750.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014840.hg.1 |
ENST00000458097.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014840.hg.1 |
ENST00000458097.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014840.hg.1 |
ENST00000458097.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014840.hg.1 |
ENST00000458097.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014840.hg.1 |
ENST00000458097.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014840.hg.1 |
ENST00000458097.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014840.hg.1 |
ENST00000458097.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014840.hg.1 |
ENST00000458097.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014840.hg.1 |
ENST00000458097.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014840.hg.1 |
ENST00000458097.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014840.hg.1 |
ENST00000458097.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014840.hg.1 |
ENST00000458097.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014840.hg.1 |
ENST00000458097.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014840.hg.1 |
ENST00000458097.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014840.hg.1 |
NR_110682.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014840.hg.1 |
NR_110682.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014840.hg.1 |
NR_110682.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014840.hg.1 |
NR_110682.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014840.hg.1 |
NR_110682.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014840.hg.1 |
NR_110682.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014840.hg.1 |
NR_110682.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100210468.hg.1 |
ENST00000425750.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100210468.hg.1 |
ENST00000458097.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100210468.hg.1 |
ENST00000458097.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100210468.hg.1 |
NR_110682.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100210468.hg.1 |
ENST00000425750.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100210468.hg.1 |
ENST00000458097.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100210468.hg.1 |
ENST00000458097.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100210468.hg.1 |
NR_110682.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100210468.hg.1 |
TC0100014840.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100014840.hg.1 |
ENST00000425750.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014840.hg.1 |
ENST00000425750.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014840.hg.1 |
ENST00000425750.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014840.hg.1 |
ENST00000425750.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014840.hg.1 |
ENST00000458097.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014840.hg.1 |
ENST00000458097.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014840.hg.1 |
ENST00000458097.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014840.hg.1 |
ENST00000458097.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014840.hg.1 |
ENST00000458097.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014840.hg.1 |
ENST00000458097.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014840.hg.1 |
ENST00000458097.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014840.hg.1 |
ENST00000458097.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014840.hg.1 |
ENST00000458097.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014840.hg.1 |
ENST00000458097.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014840.hg.1 |
ENST00000458097.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014840.hg.1 |
ENST00000458097.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014840.hg.1 |
ENST00000458097.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014840.hg.1 |
ENST00000458097.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014840.hg.1 |
NR_110682.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014840.hg.1 |
NR_110682.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014840.hg.1 |
NR_110682.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014840.hg.1 |
NR_110682.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014840.hg.1 |
NR_110682.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014840.hg.1 |
NR_110682.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014840.hg.1 |
NR_110682.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100210472.hg.1 |
ENST00000458097.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100210472.hg.1 |
ENST00000458097.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100210472.hg.1 |
NR_110682.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100210472.hg.1 |
ENST00000458097.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100210472.hg.1 |
ENST00000458097.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100210472.hg.1 |
NR_110682.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100210472.hg.1 |
TC0100014840.hg.1 |
Clariom D TC |