Assay Details
| Gene Symbol: ZNF793-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110723.1 | 2 - 3 | 441 | 96 | ||
| GenBank mRNA | AI620530.1 | - | 1 - 2 | 444 | 96 | |
| EL950706.1 | - | 2 - 3 | 323 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | ENST00000585397.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | ENST00000585397.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | ENST00000585397.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | ENST00000585890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | ENST00000585890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | ENST00000585890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | ENST00000585890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | ENST00000587060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | ENST00000587060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | ENST00000587060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | ENST00000587060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | ENST00000587060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | ENST00000588845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | ENST00000588845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | ENST00000588845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | ENST00000588845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | ENST00000588845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | ENST00000588845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | ENST00000588845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | ENST00000588845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | ENST00000588845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | ENST00000588845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | ENST00000589025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | ENST00000589025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | ENST00000589025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | ENST00000589025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | ENST00000589025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | ENST00000589025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | ENST00000589025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | ENST00000589025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | NR_110723.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | NR_110723.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | NR_110723.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | NR_110723.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | NR_110723.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | NR_110723.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | NR_110723.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010571.hg.1 | NR_110723.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900186815.hg.1 | ENST00000585890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186815.hg.1 | ENST00000588845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186815.hg.1 | ENST00000588845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186815.hg.1 | NR_110723.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900186815.hg.1 | TC1900010571.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900186815.hg.1 | NR_110723.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900186815.hg.1 | ENST00000585890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186815.hg.1 | ENST00000588845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186815.hg.1 | ENST00000588845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186819.hg.1 | ENST00000585397.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186819.hg.1 | ENST00000585890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186819.hg.1 | ENST00000588845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186819.hg.1 | ENST00000588845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186819.hg.1 | ENST00000589025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186819.hg.1 | ENST00000589025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186819.hg.1 | NR_110723.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900186819.hg.1 | TC1900010571.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900186819.hg.1 | ENST00000585397.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186819.hg.1 | ENST00000585890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186819.hg.1 | ENST00000588845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186819.hg.1 | ENST00000588845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186819.hg.1 | ENST00000589025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186819.hg.1 | ENST00000589025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186819.hg.1 | NR_110723.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900186816.hg.1 | ENST00000585890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186816.hg.1 | ENST00000588845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186816.hg.1 | ENST00000588845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186816.hg.1 | NR_110723.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900186816.hg.1 | TC1900010571.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900186816.hg.1 | ENST00000585890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186816.hg.1 | ENST00000588845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186816.hg.1 | ENST00000588845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186816.hg.1 | NR_110723.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | ENST00000585397.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | ENST00000585397.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | ENST00000585397.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | ENST00000585890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | ENST00000585890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | ENST00000585890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | ENST00000585890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | ENST00000587060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | ENST00000587060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | ENST00000587060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | ENST00000587060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | ENST00000587060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | ENST00000588845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | ENST00000588845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | ENST00000588845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | ENST00000588845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | ENST00000588845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | ENST00000588845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | ENST00000588845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | ENST00000588845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | ENST00000588845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | ENST00000588845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | ENST00000589025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | ENST00000589025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | ENST00000589025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | ENST00000589025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | ENST00000589025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | ENST00000589025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | ENST00000589025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | ENST00000589025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | NR_110723.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | NR_110723.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | NR_110723.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | NR_110723.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | NR_110723.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | NR_110723.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | NR_110723.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010571.hg.1 | NR_110723.1 | Refseq |

ZNF793-AS1
Genomic Map