| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012004.hg.1 |
ENST00000593654.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012004.hg.1 |
ENST00000593654.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012004.hg.1 |
ENST00000593654.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012004.hg.1 |
ENST00000593654.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012004.hg.1 |
ENST00000593654.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012004.hg.1 |
ENST00000593654.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012004.hg.1 |
ENST00000599259.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012004.hg.1 |
ENST00000599259.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012004.hg.1 |
ENST00000599259.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012004.hg.1 |
ENST00000599259.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012004.hg.1 |
ENST00000599259.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012004.hg.1 |
ENST00000601893.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012004.hg.1 |
ENST00000601893.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012004.hg.1 |
ENST00000601893.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012004.hg.1 |
ENST00000601893.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012004.hg.1 |
ENST00000601893.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012004.hg.1 |
ENST00000601893.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012004.hg.1 |
ENST00000601893.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012004.hg.1 |
NR_110730.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012004.hg.1 |
NR_110730.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012004.hg.1 |
NR_110730.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012004.hg.1 |
NR_110730.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012004.hg.1 |
NR_110730.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012004.hg.1 |
NR_110730.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012004.hg.1 |
NR_110730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900207166.hg.1 |
ENST00000593654.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207166.hg.1 |
ENST00000599259.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207166.hg.1 |
ENST00000601893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207166.hg.1 |
NR_110730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900207166.hg.1 |
TC1900012004.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900207166.hg.1 |
ENST00000593654.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207166.hg.1 |
ENST00000599259.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207166.hg.1 |
ENST00000601893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207166.hg.1 |
NR_110730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900207169.hg.1 |
ENST00000593654.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207169.hg.1 |
ENST00000601893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207169.hg.1 |
NR_110730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900207169.hg.1 |
TC1900012004.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900207169.hg.1 |
ENST00000593654.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207169.hg.1 |
ENST00000601893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207169.hg.1 |
NR_110730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012004.hg.1 |
ENST00000593654.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012004.hg.1 |
ENST00000593654.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012004.hg.1 |
ENST00000593654.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012004.hg.1 |
ENST00000593654.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012004.hg.1 |
ENST00000593654.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012004.hg.1 |
ENST00000593654.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012004.hg.1 |
ENST00000599259.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012004.hg.1 |
ENST00000599259.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012004.hg.1 |
ENST00000599259.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012004.hg.1 |
ENST00000599259.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012004.hg.1 |
ENST00000599259.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012004.hg.1 |
ENST00000601893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012004.hg.1 |
ENST00000601893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012004.hg.1 |
ENST00000601893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012004.hg.1 |
ENST00000601893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012004.hg.1 |
ENST00000601893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012004.hg.1 |
ENST00000601893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012004.hg.1 |
ENST00000601893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012004.hg.1 |
NR_110730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012004.hg.1 |
NR_110730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012004.hg.1 |
NR_110730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012004.hg.1 |
NR_110730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012004.hg.1 |
NR_110730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012004.hg.1 |
NR_110730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012004.hg.1 |
NR_110730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900207167.hg.1 |
ENST00000593654.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207167.hg.1 |
ENST00000601893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207167.hg.1 |
NR_110730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900207167.hg.1 |
TC1900012004.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900207167.hg.1 |
ENST00000593654.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207167.hg.1 |
ENST00000601893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207167.hg.1 |
NR_110730.1 |
Refseq |