Assay Details
| Gene Symbol: LOC101927239 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110748.1 | 3 - 4 | 471 | 123 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | acid sensing ion channel subunit 2 | ASIC2 |
ACCN, ACCN1, ASIC2a, BNC1, BNaC1, MDEG, hBNaC1 |
-21.9kb | Sense | Chr. 17 - 33013087 - 34156806 on NCBI Build GRCh38 |
| Downstream | C-C motif chemokine ligand 2 | CCL2 |
GDCF-2, HC11, HSMCR30, MCAF, MCP-1, MCP1, SCYA2, SMC-CF |
75.9kb | Anti-sense | Chr. 17 - 34255277 - 34257203 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000225823.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000225823.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000225823.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000225823.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000225823.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000225823.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000225823.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000225823.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000225823.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000225823.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000225823.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000225823.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000225823.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000225823.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000359872.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000359872.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000359872.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000359872.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000359872.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000359872.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000359872.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000359872.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000359872.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000359872.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000359872.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000359872.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000359872.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000359872.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000359872.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000359872.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000448983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000448983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000448983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000448983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000448983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000448983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000448983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000566930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000566930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000566930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000566930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000566930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000566930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000566930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000566930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000566930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000566930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000566930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000566930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000566930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000566930.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000579816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000579816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000579816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000580792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000580792.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000583395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000583395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000583395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000584724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000584724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000584724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | ENST00000584724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | NM_001094.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | NM_001094.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | NM_001094.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | NM_001094.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | NM_001094.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | NM_001094.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | NM_001094.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | NM_001094.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | NM_001094.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | NM_001094.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | NM_001094.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | NM_001094.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | NM_001094.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | NM_001094.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | NM_183377.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | NM_183377.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | NM_183377.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | NM_183377.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | NM_183377.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | NM_183377.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | NM_183377.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | NM_183377.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | NM_183377.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | NM_183377.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | NM_183377.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | NM_183377.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | NM_183377.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | NM_183377.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | NR_110748.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | NR_110748.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | NR_110748.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | NR_110748.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | NR_110748.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | NR_110748.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010371.hg.1 | NR_110748.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700181414.hg.1 | ENST00000566930.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700181414.hg.1 | ENST00000566930.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700181414.hg.1 | NR_110748.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700181414.hg.1 | TC1700010371.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700181414.hg.1 | ENST00000566930.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700181414.hg.1 | ENST00000566930.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700181414.hg.1 | NR_110748.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000225823.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000225823.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000225823.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000225823.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000225823.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000225823.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000225823.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000225823.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000225823.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000225823.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000225823.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000225823.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000225823.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000225823.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000359872.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000359872.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000359872.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000359872.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000359872.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000359872.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000359872.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000359872.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000359872.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000359872.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000359872.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000359872.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000359872.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000359872.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000359872.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000359872.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000448983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000448983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000448983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000448983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000448983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000448983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000448983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000566930.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000566930.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000566930.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000566930.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000566930.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000566930.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000566930.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000566930.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000566930.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000566930.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000566930.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000566930.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000566930.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000566930.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000579816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000579816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000579816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000580792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000580792.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000583395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000583395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000583395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000584724.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000584724.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000584724.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | ENST00000584724.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | NM_001094.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | NM_001094.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | NM_001094.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | NM_001094.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | NM_001094.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | NM_001094.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | NM_001094.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | NM_001094.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | NM_001094.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | NM_001094.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | NM_001094.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | NM_001094.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | NM_001094.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | NM_001094.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | NM_183377.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | NM_183377.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | NM_183377.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | NM_183377.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | NM_183377.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | NM_183377.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | NM_183377.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | NM_183377.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | NM_183377.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | NM_183377.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | NM_183377.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | NM_183377.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | NM_183377.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | NM_183377.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | NR_110748.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | NR_110748.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | NR_110748.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | NR_110748.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | NR_110748.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | NR_110748.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010371.hg.1 | NR_110748.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700181413.hg.1 | ENST00000566930.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700181413.hg.1 | ENST00000566930.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700181413.hg.1 | NR_110748.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700181413.hg.1 | TC1700010371.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700181413.hg.1 | ENST00000566930.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700181413.hg.1 | ENST00000566930.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700181413.hg.1 | NR_110748.1 | Refseq |

LOC101927239
Genomic Map