| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006558.hg.1 |
ENST00000578427.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006558.hg.1 |
ENST00000578427.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006558.hg.1 |
ENST00000578427.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006558.hg.1 |
NR_110765.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006558.hg.1 |
NR_110765.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006558.hg.1 |
NR_110765.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800144926.hg.1 |
ENST00000578427.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800144926.hg.1 |
NR_110765.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800144926.hg.1 |
TC1800006558.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1800144926.hg.1 |
ENST00000578427.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800144926.hg.1 |
NR_110765.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800144922.hg.1 |
ENST00000578427.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800144922.hg.1 |
NR_110765.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800144922.hg.1 |
TC1800006558.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1800144922.hg.1 |
ENST00000578427.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800144922.hg.1 |
NR_110765.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800006558.hg.1 |
ENST00000578427.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006558.hg.1 |
ENST00000578427.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006558.hg.1 |
ENST00000578427.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006558.hg.1 |
NR_110765.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800006558.hg.1 |
NR_110765.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800006558.hg.1 |
NR_110765.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800144925.hg.1 |
ENST00000578427.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800144925.hg.1 |
NR_110765.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800144925.hg.1 |
TC1800006558.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1800144925.hg.1 |
ENST00000578427.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800144925.hg.1 |
NR_110765.1 |
Refseq |