| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008391.hg.1 |
ENST00000578119.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008391.hg.1 |
ENST00000578119.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008391.hg.1 |
ENST00000578119.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008391.hg.1 |
ENST00000578477.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008391.hg.1 |
ENST00000578477.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008391.hg.1 |
ENST00000578477.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008391.hg.1 |
ENST00000578477.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008391.hg.1 |
ENST00000578477.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008391.hg.1 |
ENST00000578477.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008391.hg.1 |
ENST00000578477.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008391.hg.1 |
ENST00000578477.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008391.hg.1 |
ENST00000578477.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008391.hg.1 |
ENST00000578477.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008391.hg.1 |
ENST00000578477.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008391.hg.1 |
ENST00000578477.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008391.hg.1 |
ENST00000581452.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008391.hg.1 |
ENST00000581452.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008391.hg.1 |
ENST00000581452.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008391.hg.1 |
ENST00000581452.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008391.hg.1 |
ENST00000581856.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008391.hg.1 |
ENST00000581856.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008391.hg.1 |
ENST00000581856.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008391.hg.1 |
ENST00000581856.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008391.hg.1 |
ENST00000581856.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008391.hg.1 |
ENST00000581856.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008391.hg.1 |
ENST00000581856.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008391.hg.1 |
ENST00000581856.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008391.hg.1 |
NR_110788.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008391.hg.1 |
NR_110788.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008391.hg.1 |
NR_110788.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008391.hg.1 |
NR_110788.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008391.hg.1 |
NR_110788.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008391.hg.1 |
NR_110789.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008391.hg.1 |
NR_110789.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008391.hg.1 |
NR_110789.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800157181.hg.1 |
ENST00000578119.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800157181.hg.1 |
ENST00000578477.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800157181.hg.1 |
ENST00000578477.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800157181.hg.1 |
ENST00000581856.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800157181.hg.1 |
ENST00000581856.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800157181.hg.1 |
NR_110788.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800157181.hg.1 |
NR_110789.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800157181.hg.1 |
TC1800008391.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1800157181.hg.1 |
ENST00000578119.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800157181.hg.1 |
ENST00000578477.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800157181.hg.1 |
ENST00000578477.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800157181.hg.1 |
ENST00000581856.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800157181.hg.1 |
ENST00000581856.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800157181.hg.1 |
NR_110788.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800157181.hg.1 |
NR_110789.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800157182.hg.1 |
ENST00000578119.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800157182.hg.1 |
NR_110789.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800157182.hg.1 |
TC1800008391.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1800157182.hg.1 |
ENST00000578119.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800157182.hg.1 |
NR_110789.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800157179.hg.1 |
ENST00000578119.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800157179.hg.1 |
ENST00000581856.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800157179.hg.1 |
ENST00000581856.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800157179.hg.1 |
NR_110788.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800157179.hg.1 |
NR_110789.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800157179.hg.1 |
TC1800008391.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1800157179.hg.1 |
ENST00000578119.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800157179.hg.1 |
ENST00000581856.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800157179.hg.1 |
ENST00000581856.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800157179.hg.1 |
NR_110788.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800157179.hg.1 |
NR_110789.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008391.hg.1 |
ENST00000578119.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008391.hg.1 |
ENST00000578119.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008391.hg.1 |
ENST00000578119.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008391.hg.1 |
ENST00000578477.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008391.hg.1 |
ENST00000578477.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008391.hg.1 |
ENST00000578477.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008391.hg.1 |
ENST00000578477.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008391.hg.1 |
ENST00000578477.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008391.hg.1 |
ENST00000578477.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008391.hg.1 |
ENST00000578477.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008391.hg.1 |
ENST00000578477.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008391.hg.1 |
ENST00000578477.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008391.hg.1 |
ENST00000578477.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008391.hg.1 |
ENST00000578477.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008391.hg.1 |
ENST00000578477.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008391.hg.1 |
ENST00000581452.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008391.hg.1 |
ENST00000581452.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008391.hg.1 |
ENST00000581452.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008391.hg.1 |
ENST00000581452.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008391.hg.1 |
ENST00000581856.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008391.hg.1 |
ENST00000581856.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008391.hg.1 |
ENST00000581856.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008391.hg.1 |
ENST00000581856.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008391.hg.1 |
ENST00000581856.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008391.hg.1 |
ENST00000581856.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008391.hg.1 |
ENST00000581856.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008391.hg.1 |
ENST00000581856.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008391.hg.1 |
NR_110788.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008391.hg.1 |
NR_110788.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008391.hg.1 |
NR_110788.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008391.hg.1 |
NR_110788.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008391.hg.1 |
NR_110788.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008391.hg.1 |
NR_110789.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008391.hg.1 |
NR_110789.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008391.hg.1 |
NR_110789.1 |
Refseq |