Assay Details
| Gene Symbol: LINC01478 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110792.1 | 1 - 2 | 238 | 90 | ||
| GenBank mRNA | BC051727.1 | - | 1 - 2 | 238 | 90 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | synaptotagmin 4 | SYT4 | HsT1192 | -1.2mb | Sense | Chr. 18 - 43267892 - 43277650 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101927943 | LOC101927943 | 145.2kb | Sense | Chr. 18 - 44676927 - 44679717 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LINC01478
Genomic Map