| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009273.hg.1 |
ENST00000587877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009273.hg.1 |
ENST00000587877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009273.hg.1 |
ENST00000587877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009273.hg.1 |
ENST00000587877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009273.hg.1 |
ENST00000587877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009273.hg.1 |
ENST00000587877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009273.hg.1 |
ENST00000587877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009273.hg.1 |
ENST00000587877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009273.hg.1 |
ENST00000591487.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009273.hg.1 |
ENST00000591487.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009273.hg.1 |
ENST00000591487.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009273.hg.1 |
ENST00000591487.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009273.hg.1 |
NR_110792.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009273.hg.1 |
NR_110792.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009273.hg.1 |
NR_110792.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009273.hg.1 |
NR_110792.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009273.hg.1 |
NR_110792.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009273.hg.1 |
NR_110792.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009273.hg.1 |
NR_110792.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009273.hg.1 |
NR_110792.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800009273.hg.1 |
ENST00000587877.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009273.hg.1 |
ENST00000587877.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009273.hg.1 |
ENST00000587877.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009273.hg.1 |
ENST00000587877.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009273.hg.1 |
ENST00000587877.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009273.hg.1 |
ENST00000587877.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009273.hg.1 |
ENST00000587877.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009273.hg.1 |
ENST00000587877.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009273.hg.1 |
ENST00000591487.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009273.hg.1 |
ENST00000591487.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009273.hg.1 |
ENST00000591487.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009273.hg.1 |
ENST00000591487.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009273.hg.1 |
NR_110792.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800009273.hg.1 |
NR_110792.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800009273.hg.1 |
NR_110792.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800009273.hg.1 |
NR_110792.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800009273.hg.1 |
NR_110792.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800009273.hg.1 |
NR_110792.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800009273.hg.1 |
NR_110792.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800009273.hg.1 |
NR_110792.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800163987.hg.1 |
ENST00000587877.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800163987.hg.1 |
NR_110792.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800163987.hg.1 |
TC1800009273.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1800163987.hg.1 |
ENST00000587877.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800163987.hg.1 |
NR_110792.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800163988.hg.1 |
ENST00000587877.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800163988.hg.1 |
NR_110792.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800163988.hg.1 |
TC1800009273.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1800163988.hg.1 |
ENST00000587877.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800163988.hg.1 |
NR_110792.1 |
Refseq |