Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008434.hg.1 |
ENST00000578022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008434.hg.1 |
ENST00000578022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008434.hg.1 |
ENST00000578022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008434.hg.1 |
ENST00000578022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008434.hg.1 |
ENST00000578022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008434.hg.1 |
ENST00000578022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008434.hg.1 |
ENST00000578022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008434.hg.1 |
ENST00000580589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008434.hg.1 |
ENST00000580589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008434.hg.1 |
ENST00000580589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008434.hg.1 |
ENST00000580589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008434.hg.1 |
ENST00000580589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008434.hg.1 |
ENST00000580589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008434.hg.1 |
ENST00000580589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008434.hg.1 |
ENST00000580589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008434.hg.1 |
ENST00000580589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008434.hg.1 |
ENST00000580589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008434.hg.1 |
ENST00000580769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008434.hg.1 |
ENST00000580769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008434.hg.1 |
ENST00000580769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008434.hg.1 |
ENST00000580769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008434.hg.1 |
NR_110810.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008434.hg.1 |
NR_110810.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008434.hg.1 |
NR_110810.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008434.hg.1 |
NR_110810.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008434.hg.1 |
NR_110810.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008434.hg.1 |
ENST00000578022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008434.hg.1 |
ENST00000578022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008434.hg.1 |
ENST00000578022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008434.hg.1 |
ENST00000578022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008434.hg.1 |
ENST00000578022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008434.hg.1 |
ENST00000578022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008434.hg.1 |
ENST00000578022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008434.hg.1 |
ENST00000580589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008434.hg.1 |
ENST00000580589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008434.hg.1 |
ENST00000580589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008434.hg.1 |
ENST00000580589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008434.hg.1 |
ENST00000580589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008434.hg.1 |
ENST00000580589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008434.hg.1 |
ENST00000580589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008434.hg.1 |
ENST00000580589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008434.hg.1 |
ENST00000580589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008434.hg.1 |
ENST00000580589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008434.hg.1 |
ENST00000580769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008434.hg.1 |
ENST00000580769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008434.hg.1 |
ENST00000580769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008434.hg.1 |
ENST00000580769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008434.hg.1 |
NR_110810.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008434.hg.1 |
NR_110810.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008434.hg.1 |
NR_110810.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008434.hg.1 |
NR_110810.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008434.hg.1 |
NR_110810.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700162745.hg.1 |
ENST00000578022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162745.hg.1 |
ENST00000580589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162745.hg.1 |
ENST00000580589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162745.hg.1 |
ENST00000580769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162745.hg.1 |
NR_110810.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700162745.hg.1 |
ENST00000578022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162745.hg.1 |
ENST00000580589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162745.hg.1 |
ENST00000580589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162745.hg.1 |
ENST00000580769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162745.hg.1 |
NR_110810.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700162745.hg.1 |
TC1700008434.hg.1 |
Clariom D TC |