Assay Details
| Gene Symbol: HP09053 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110821.1 | 2 - 3 | 173 | 103 | ||
| GenBank mRNA | AB593151.1 | - | 2 - 3 | 173 | 103 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cms1 ribosomal small subunit homolog (yeast) | CMSS1 | C3orf26 | Anti-sense | Chr. 3 - 99817834 - 100178632 on NCBI Build GRCh38 | |
| Downstream | filamin A interacting protein 1 like | FILIP1L |
DOC-1, DOC1, GIP130, GIP90 |
14.2kb | Sense | Chr. 3 - 99833144 - 100114513 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

HP09053
Genomic Map