| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
ENST00000418665.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
ENST00000418665.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
ENST00000418665.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
ENST00000418665.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
ENST00000418665.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
ENST00000418665.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
ENST00000418665.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
ENST00000418665.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
ENST00000418665.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
ENST00000433489.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
ENST00000433489.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
ENST00000433489.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
ENST00000433489.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
ENST00000433489.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
ENST00000433489.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
ENST00000433489.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
ENST00000433489.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
ENST00000433489.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
ENST00000433489.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
ENST00000433489.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
ENST00000433489.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
ENST00000433489.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
ENST00000433489.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
ENST00000440924.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
ENST00000440924.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
ENST00000440924.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
ENST00000440924.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
ENST00000440924.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
ENST00000440924.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
ENST00000440924.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
ENST00000440924.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
ENST00000440924.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
ENST00000440924.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
NR_110871.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
NR_110871.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
NR_110871.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
NR_110871.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
NR_110871.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
NR_110871.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
NR_110871.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
NR_110871.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
NR_110871.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
NR_110872.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
NR_110872.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
NR_110872.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
NR_110872.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
NR_110872.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013812.hg.1 |
NR_110872.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
ENST00000418665.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
ENST00000418665.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
ENST00000418665.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
ENST00000418665.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
ENST00000418665.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
ENST00000418665.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
ENST00000418665.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
ENST00000418665.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
ENST00000418665.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
ENST00000433489.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
ENST00000433489.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
ENST00000433489.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
ENST00000433489.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
ENST00000433489.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
ENST00000433489.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
ENST00000433489.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
ENST00000433489.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
ENST00000433489.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
ENST00000433489.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
ENST00000433489.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
ENST00000433489.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
ENST00000433489.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
ENST00000433489.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
ENST00000440924.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
ENST00000440924.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
ENST00000440924.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
ENST00000440924.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
ENST00000440924.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
ENST00000440924.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
ENST00000440924.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
ENST00000440924.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
ENST00000440924.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
ENST00000440924.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
NR_110871.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
NR_110871.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
NR_110871.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
NR_110871.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
NR_110871.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
NR_110871.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
NR_110871.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
NR_110871.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
NR_110871.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
NR_110872.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
NR_110872.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
NR_110872.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
NR_110872.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
NR_110872.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013812.hg.1 |
NR_110872.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600195298.hg.1 |
ENST00000418665.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600195298.hg.1 |
NR_110871.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600195298.hg.1 |
NR_110872.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600195298.hg.1 |
ENST00000418665.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600195298.hg.1 |
NR_110871.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600195298.hg.1 |
NR_110872.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600195298.hg.1 |
TC0600013812.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600195297.hg.1 |
ENST00000418665.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600195297.hg.1 |
NR_110871.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600195297.hg.1 |
NR_110872.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600195297.hg.1 |
ENST00000418665.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600195297.hg.1 |
NR_110871.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600195297.hg.1 |
NR_110872.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600195297.hg.1 |
TC0600013812.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600195293.hg.1 |
NR_110872.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600195293.hg.1 |
NR_110872.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600195293.hg.1 |
TC0600013812.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600195296.hg.1 |
ENST00000418665.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600195296.hg.1 |
NR_110871.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600195296.hg.1 |
NR_110872.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600195296.hg.1 |
ENST00000418665.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600195296.hg.1 |
NR_110871.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600195296.hg.1 |
NR_110872.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600195296.hg.1 |
TC0600013812.hg.1 |
Clariom D TC |