Assay Details
| Gene Symbol: FAM65C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110890.1 | 1 - 2 | 591 | 62 | ||
| GenBank mRNA | AK295781.1 | - | 1 - 2 | 282 | 62 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100506175 | LOC100506175 | Anti-sense | Chr. 20 - 50645471 - 50660291 on NCBI Build GRCh38 | ||
| Downstream | par-6 family cell polarity regulator beta | PARD6B | PAR6B | 40.0kb | Anti-sense | Chr. 20 - 50731544 - 50753741 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK295781.1 transcripts.
Set Membership: |
|

FAM65C
Genomic Map