Assay Details
| Gene Symbol: ZNF213-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110900.1 | 5 - 6 | 686 | 71 | ||
| GenBank mRNA | AI934565.1 | - | 1 - 2 | 71 | ||
| BG752676.1 | - | 5 - 6 | 641 | 70 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 205 | ZNF205 |
RhitH, ZNF210, Zfp13 |
-5.0kb | Anti-sense | Chr. 16 - 3112562 - 3120517 on NCBI Build GRCh38 |
| Downstream | zinc finger protein 213 | ZNF213 |
CR53, ZKSCAN21, ZSCAN53 |
5.7kb | Anti-sense | Chr. 16 - 3135056 - 3142804 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000570901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000570901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000570901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000570901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000573414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000573414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000573414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000575089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000575089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000575089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000575089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000575089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000576590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000576590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000576590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000576590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000577163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000577163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000577163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000577163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | ENST00000577163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600192369.hg.1 | ENST00000570901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192369.hg.1 | ENST00000573414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192369.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600192369.hg.1 | TC1600011471.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600192369.hg.1 | ENST00000570901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192369.hg.1 | ENST00000573414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192369.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600192368.hg.1 | ENST00000570901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192368.hg.1 | ENST00000573414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192368.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600192368.hg.1 | TC1600011471.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600192368.hg.1 | ENST00000570901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192368.hg.1 | ENST00000573414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192368.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000570901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000570901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000570901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000570901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000571963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000573414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000573414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000573414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000573447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000575089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000575089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000575089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000575089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000575089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000576590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000576590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000576590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000576590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000577163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000577163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000577163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000577163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | ENST00000577163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011471.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600192367.hg.1 | ENST00000570901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192367.hg.1 | NR_110900.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600192367.hg.1 | TC1600011471.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600192367.hg.1 | ENST00000570901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600192367.hg.1 | NR_110900.1 | Refseq |

ZNF213-AS1
Genomic Map