| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007733.hg.1 |
ENST00000564705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007733.hg.1 |
ENST00000564705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007733.hg.1 |
ENST00000564705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007733.hg.1 |
ENST00000564705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007733.hg.1 |
ENST00000564705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007733.hg.1 |
ENST00000564705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007733.hg.1 |
ENST00000564705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007733.hg.1 |
ENST00000564705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007733.hg.1 |
ENST00000564705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007733.hg.1 |
ENST00000564705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007733.hg.1 |
ENST00000564705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007733.hg.1 |
ENST00000564705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007733.hg.1 |
ENST00000565694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007733.hg.1 |
ENST00000565694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007733.hg.1 |
ENST00000565694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007733.hg.1 |
ENST00000565694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007733.hg.1 |
ENST00000565694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007733.hg.1 |
ENST00000565694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007733.hg.1 |
NR_110903.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007733.hg.1 |
NR_110903.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007733.hg.1 |
NR_110903.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007733.hg.1 |
NR_110903.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007733.hg.1 |
NR_110903.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007733.hg.1 |
NR_110903.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007733.hg.1 |
NR_110903.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007733.hg.1 |
NR_110903.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007733.hg.1 |
NR_110903.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007733.hg.1 |
NR_110904.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007733.hg.1 |
NR_110904.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007733.hg.1 |
NR_110904.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007733.hg.1 |
NR_110904.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007733.hg.1 |
NR_110904.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007733.hg.1 |
NR_110904.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007733.hg.1 |
NR_110904.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007733.hg.1 |
NR_110904.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600156402.hg.1 |
ENST00000564705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600156402.hg.1 |
ENST00000564705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600156402.hg.1 |
ENST00000565694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600156402.hg.1 |
NR_110903.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600156402.hg.1 |
NR_110904.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600156402.hg.1 |
TC1600007733.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600156402.hg.1 |
ENST00000564705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600156402.hg.1 |
ENST00000564705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600156402.hg.1 |
ENST00000565694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600156402.hg.1 |
NR_110903.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600156402.hg.1 |
NR_110904.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007733.hg.1 |
ENST00000564705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007733.hg.1 |
ENST00000564705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007733.hg.1 |
ENST00000564705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007733.hg.1 |
ENST00000564705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007733.hg.1 |
ENST00000564705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007733.hg.1 |
ENST00000564705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007733.hg.1 |
ENST00000564705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007733.hg.1 |
ENST00000564705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007733.hg.1 |
ENST00000564705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007733.hg.1 |
ENST00000564705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007733.hg.1 |
ENST00000564705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007733.hg.1 |
ENST00000564705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007733.hg.1 |
ENST00000565694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007733.hg.1 |
ENST00000565694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007733.hg.1 |
ENST00000565694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007733.hg.1 |
ENST00000565694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007733.hg.1 |
ENST00000565694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007733.hg.1 |
ENST00000565694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007733.hg.1 |
NR_110903.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007733.hg.1 |
NR_110903.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007733.hg.1 |
NR_110903.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007733.hg.1 |
NR_110903.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007733.hg.1 |
NR_110903.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007733.hg.1 |
NR_110903.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007733.hg.1 |
NR_110903.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007733.hg.1 |
NR_110903.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007733.hg.1 |
NR_110903.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007733.hg.1 |
NR_110904.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007733.hg.1 |
NR_110904.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007733.hg.1 |
NR_110904.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007733.hg.1 |
NR_110904.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007733.hg.1 |
NR_110904.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007733.hg.1 |
NR_110904.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007733.hg.1 |
NR_110904.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007733.hg.1 |
NR_110904.1 |
Refseq |