Assay Details
| Gene Symbol: LOC101929613 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110948.1 | 4 - 5 | 159 | 74 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 514 | LINC00514 | LA16c-380H5.1 | -5.8kb | Sense | Chr. 16 - 2989054 - 2994509 on NCBI Build GRCh38 |
| Downstream | claudin 9 | CLDN9 | 11.6kb | Sense | Chr. 16 - 3012456 - 3014505 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC101929613
Genomic Map