| Array Name | 
									Probe IDs | 
									Targets | 
									Target Type | 
								
	
								
									
										
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1500007434.hg.1 | 
												ENST00000558104.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1500007434.hg.1 | 
												ENST00000558104.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1500007434.hg.1 | 
												ENST00000558104.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1500007434.hg.1 | 
												ENST00000558104.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1500007434.hg.1 | 
												ENST00000561413.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1500007434.hg.1 | 
												ENST00000561413.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1500007434.hg.1 | 
												ENST00000561413.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1500007434.hg.1 | 
												NR_120318.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1500007434.hg.1 | 
												NR_120318.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1500007434.hg.1 | 
												NR_120318.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1500007434.hg.1 | 
												NR_120318.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1500007434.hg.1 | 
												NR_120318.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1500007434.hg.1 | 
												NR_120318.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1500007434.hg.1 | 
												NR_120319.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1500007434.hg.1 | 
												NR_120319.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1500007434.hg.1 | 
												NR_120319.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1500007434.hg.1 | 
												NR_120319.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1500007434.hg.1 | 
												NR_120319.1 | 
												Refseq | 
												
											
										
										
									
								
									
										
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1500007434.hg.1 | 
												ENST00000558104.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1500007434.hg.1 | 
												ENST00000558104.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1500007434.hg.1 | 
												ENST00000558104.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1500007434.hg.1 | 
												ENST00000558104.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1500007434.hg.1 | 
												ENST00000561413.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1500007434.hg.1 | 
												ENST00000561413.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1500007434.hg.1 | 
												ENST00000561413.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1500007434.hg.1 | 
												NR_120318.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1500007434.hg.1 | 
												NR_120318.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1500007434.hg.1 | 
												NR_120318.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1500007434.hg.1 | 
												NR_120318.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1500007434.hg.1 | 
												NR_120318.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1500007434.hg.1 | 
												NR_120318.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1500007434.hg.1 | 
												NR_120319.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1500007434.hg.1 | 
												NR_120319.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1500007434.hg.1 | 
												NR_120319.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1500007434.hg.1 | 
												NR_120319.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1500007434.hg.1 | 
												NR_120319.1 | 
												Refseq | 
												
											
										
										
									
								
									
										
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1500152849.hg.1 | 
												ENST00000561413.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1500152849.hg.1 | 
												NR_120318.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1500152849.hg.1 | 
												ENST00000561413.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1500152849.hg.1 | 
												NR_120318.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1500152849.hg.1 | 
												TC1500007434.hg.1 | 
												Clariom D TC |